Pentafluorophenyl trifluoroacetate 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Trifluoroacetic Acid Pentafluorophenyl Ester, Pentafluorphenyl trifluoracetate

PC8848-100G
APOSPC8848-100GEA 333 EUR
APOSPC8848-100G
Pentafluorophenyl trifluoroacetate 98%
Pentafluorophenyl trifluoroacetate
Formule: C₈F₈O₂
Kookpunt: 122…123 °C
Smeltpunt: 3 °C
Dichtheid: 1,63 g/cm³ (25 °C)
Vlampunt: 52 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00134438
CAS nummer: 14533-84-7
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR