2-Hydrazino-4-(trifluoromethyl)pyridine 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC8988-100MG PC8988-1G PC8988-250MG
APOSPC8988-100MGEA 38 EUR
APOSPC8988-100MG APOSPC8988-1G APOSPC8988-250MG
2-Hydrazino-4-(trifluoromethyl)pyridine 95%
2-Hydrazino-4-(trifluoromethyl)pyridine
Formule: C₆H₆F₃N₃
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD08056340
CAS nummer: 89570-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR