1-(5-Fluoropyridin-2-yl)ethanone 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC900560-1G PC900560-250MG
APOSPC900560-1GEA 144 EUR
APOSPC900560-1G APOSPC900560-250MG
1-(5-Fluoropyridin-2-yl)ethanone 98%
1-(5-Fluoropyridin-2-yl)ethanone
Formule: C₇H₆FNO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD10698609
CAS nummer: 915720-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR