5-Fluoro-2-methoxy-3-nitropyridine 96%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC900607-100MG PC900607-1G PC900607-250MG
APOSPC900607-100MGEA 256 EUR
APOSPC900607-100MG APOSPC900607-1G APOSPC900607-250MG
5-Fluoro-2-methoxy-3-nitropyridine 96%
5-Fluoro-2-methoxy-3-nitropyridine
Formule: C₆H₅FN₂O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD18260977
CAS nummer: 1211534-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR