5-Chloro-3-fluoro-2-hydroxypyridine 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 5-Chloro-3-fluoropyridin-2-ol

PC900618-5G
APOSPC900618-5GEA 141 EUR
APOSPC900618-5G
5-Chloro-3-fluoro-2-hydroxypyridine 98%
5-Chloro-3-fluoro-2-hydroxypyridine
Formule: C₅H₃ClFNO
MW: 147.535 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD07375044
CAS nummer: 514797-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR