2,3-Difluoro-N-methyl-6-nitroaniline 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC902355-100G PC902355-25G PC902355-5G
APOSPC902355-100GEA 636 EUR
APOSPC902355-100G APOSPC902355-25G APOSPC902355-5G
2,3-Difluoro-N-methyl-6-nitroaniline 97%
2,3-Difluoro-N-methyl-6-nitroaniline
Formule: C₇H₆F₂N₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD16787128
CAS nummer: 170432-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR