2-Bromo-4-fluorophenyl isothiocyanate, Technical Grade

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC9026-5G PC9026-25G
APOSPC9026-5GEA 215 EUR
APOSPC9026-5G APOSPC9026-25G
2-Bromo-4-fluorophenyl isothiocyanate, Technical Grade
2-Bromo-4-fluorophenyl isothiocyanate
Formule: C₇H₃BrFNS
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00673194
CAS nummer: 183995-72-4
UN: 2922
ADR: 8+6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR