Ethyl-3-amino-4-fluorobenzoate 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC902645-25G PC902645-5G
APOSPC902645-25GEA 238 EUR
APOSPC902645-25G APOSPC902645-5G
Ethyl-3-amino-4-fluorobenzoate 95%
Ethyl-3-amino-4-fluorobenzoate
Formule: C₉H₁₀FNO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03840487
CAS nummer: 455-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR