2-Amino-4,6-difluoropyrimidine 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 4,6-Difluoropyrimidin-2-amine

PC902684-1G PC902684-5G
APOSPC902684-1GEA 106 EUR
APOSPC902684-1G APOSPC902684-5G
2-Amino-4,6-difluoropyrimidine 95%
2-Amino-4,6-difluoropyrimidine
Formule: C₄H₃F₂N₃
MW: 131.084 g/mol
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00233527
CAS nummer: 675-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR