5-Methoxy-2-(trifluoromethyl)benzoic acid 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC903941-10G PC903941-1G PC903941-5G
APOSPC903941-10GEA 1060 EUR
APOSPC903941-10G APOSPC903941-1G APOSPC903941-5G
5-Methoxy-2-(trifluoromethyl)benzoic acid 98%
5-Methoxy-2-(trifluoromethyl)benzoic acid
Formule: C₉H₇F₃O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD13185683
CAS nummer: 716-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR