(3,5-Difluoro-4-nitrophenyl)methanol 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC904722-1G PC904722-250MG
APOSPC904722-1GEA 89.2 EUR
APOSPC904722-1G APOSPC904722-250MG
(3,5-Difluoro-4-nitrophenyl)methanol 95%
(3,5-Difluoro-4-nitrophenyl)methanol
Formule: C₇H₅F₂NO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD18633079
CAS nummer: 1123172-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR