1-(2-Chloro-5-fluoropyridin-3-yl)ethanone 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC904959-1G PC904959-250MG
APOSPC904959-1GEA 110 EUR
APOSPC904959-1G APOSPC904959-250MG
1-(2-Chloro-5-fluoropyridin-3-yl)ethanone 95%
1-(2-Chloro-5-fluoropyridin-3-yl)ethanone
Formule: C₇H₅ClFNO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD13563063
CAS nummer: 1203499-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR