5-(4-Fluorophenyl)isoxazol-3-amine 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC905317-1G PC905317-5G
APOSPC905317-1GEA 54.1 EUR
APOSPC905317-1G APOSPC905317-5G
5-(4-Fluorophenyl)isoxazol-3-amine 95%
5-(4-Fluorophenyl)isoxazol-3-amine
Formule: C₉H₇FN₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08447045
CAS nummer: 925005-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR