3-(1,1,2,2-Tetrafluoroethoxy)benzoic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC9058-100G PC9058-25G
APOSPC9058-100GEA 0 EUR
APOSPC9058-100G APOSPC9058-25G
3-(1,1,2,2-Tetrafluoroethoxy)benzoic acid
3-(1,1,2,2-Tetrafluoroethoxy)benzoic acid
Formule: C₉H₆F₄O₃
Smeltpunt: 127…128 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00155925
CAS nummer: 70126-48-6

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR