cis-4-(Trifluoromethyl)cyclohexanecarboxylic acid 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC906397-1G PC906397-250MG PC906397-5G
APOSPC906397-1GEA 172 EUR
APOSPC906397-1G APOSPC906397-250MG APOSPC906397-5G
cis-4-(Trifluoromethyl)cyclohexanecarboxylic acid 95%
cis-4-(Trifluoromethyl)cyclohexanecarboxylic acid
Formule: C₈H₁₁F₃O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09037361
CAS nummer: 1202578-27-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR