2-Amino-4-fluoro-5-methylbenzonitrile 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC906695-1G
APOSPC906695-1GEA 97.9 EUR
APOSPC906695-1G
2-Amino-4-fluoro-5-methylbenzonitrile 95%
2-Amino-4-fluoro-5-methylbenzonitrile
Formule: C₈H₇FN₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD15529743
CAS nummer: 1357942-79-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR