6-Fluoro-o-toluonitrile 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC9287-1G PC9287-5G PC9287-10G
APOSPC9287-1GEA 22 EUR
APOSPC9287-1G APOSPC9287-5G APOSPC9287-10G
6-Fluoro-o-toluonitrile 99%
6-Fluoro-o-toluonitrile
Formule: C₈H₆FN
Smeltpunt: 43…45 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06660277
CAS nummer: 198633-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR