5-Methoxy-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbonitrile

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC9388-1G
APOSPC9388-1GEA 402 EUR
APOSPC9388-1G
5-Methoxy-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbonitrile
5-Methoxy-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbonitrile
Formule: C₇H₆F₃N₃O
Smeltpunt: 36…37 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01935988
CAS nummer: 321848-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR