Methyl-4-[2-amino-4-(trifluoromethyl)anilino]butanoate 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC9391-1G PC9391-500MG PC9391-5G
APOSPC9391-1GEA 537 EUR
APOSPC9391-1G APOSPC9391-500MG APOSPC9391-5G
Methyl-4-[2-amino-4-(trifluoromethyl)anilino]butanoate 97%
Methyl-4-[2-amino-4-(trifluoromethyl)anilino]butanoate
Formule: C₁₂H₁₅F₃N₂O₂
Smeltpunt: 91…92 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00793102
CAS nummer: 339101-34-7

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR