4-{[3-(Trifluoromethyl)phenyl]sulphanyl}aniline

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC9498-1G PC9498-500MG PC9498-5G
APOSPC9498-1GEA 219 EUR
APOSPC9498-1G APOSPC9498-500MG APOSPC9498-5G
4-{[3-(Trifluoromethyl)phenyl]sulphanyl}aniline
4-{[3-(Trifluoromethyl)phenyl]sulphanyl}aniline
Formule: C₁₃H₁₀F₃NS
Smeltpunt: 64…69 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00172345
CAS nummer: 339104-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR