2-(3-Chloro-4-fluorophenyl)ethanol 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC9510-1G PC9510-5G PC9510-25G
APOSPC9510-1GEA 55.5 EUR
APOSPC9510-1G APOSPC9510-5G APOSPC9510-25G
2-(3-Chloro-4-fluorophenyl)ethanol 97%
2-(3-Chloro-4-fluorophenyl)ethanol
Formule: C₈H₈ClFO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06201121
CAS nummer: 340825-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR