4-Chloro-3-(trifluoromethyl)phenacyl bromide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC9516-1G PC9516-5G PC9516-25G
APOSPC9516-1GEA 93.5 EUR
APOSPC9516-1G APOSPC9516-5G APOSPC9516-25G
4-Chloro-3-(trifluoromethyl)phenacyl bromide
4-Chloro-3-(trifluoromethyl)phenacyl bromide
Formule: C₉H₅BrClF₃O
Smeltpunt: 36…38 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03094558
CAS nummer: 630404-09-0
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR