3-Methoxy-5-(trifluoromethyl)benzonitrile 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC9552-1G PC9552-25G PC9552-5G
APOSPC9552-1GEA 22 EUR
APOSPC9552-1G APOSPC9552-25G APOSPC9552-5G
3-Methoxy-5-(trifluoromethyl)benzonitrile 97%
3-Methoxy-5-(trifluoromethyl)benzonitrile
Formule: C₉H₆F₃NO
Smeltpunt: 38 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08458145
CAS nummer: 868167-61-7
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR