5-Chloro-2-fluorophenyl isothiocyanate 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC9564-1G PC9564-5G PC9564-250MG
APOSPC9564-1GEA 73.2 EUR
APOSPC9564-1G APOSPC9564-5G APOSPC9564-250MG
5-Chloro-2-fluorophenyl isothiocyanate 97%
5-Chloro-2-fluorophenyl isothiocyanate
Formule: C₇H₃ClFNS
Bewaar temperatuur: Kamertemperatuur
CAS nummer: 247170-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR