Perfluoropentanoyl chloride 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC9708-1G PC9708-25G PC9708-5G
APOSPC9708-1GEA 199 EUR
APOSPC9708-1G APOSPC9708-25G APOSPC9708-5G
Perfluoropentanoyl chloride 97%
Perfluoropentanoyl chloride
Formule: C₅ClF₉O
Kookpunt: 67,5…68 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00155890
CAS nummer: 375-60-0
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR