Tritylium tetrafluoroborate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC9789-25G PC9789-5G
APOSPC9789-25GEA 191 EUR
APOSPC9789-25G APOSPC9789-5G
Tritylium tetrafluoroborate
Tritylium tetrafluoroborate
Formule: C₁₉H₁₅BF₄
Smeltpunt: 205…215 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00013120
CAS nummer: 341-02-6
EINECS: 206-433-3
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR