Terrestrial (Cypripedium) orchid medium
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PMT839A-10X1L PMT839B-10L PMT839C-50L
APOSPMT839A-10X1LEA 47.1 EUR
APOSPMT839A-10X1L APOSPMT839B-10L APOSPMT839C-50L
Terrestrial (Cypripedium) orchid medium
Celcultuurmedia
Contains 400 mg/L calcium nitrate and 400 mg/L Casein; WITHOUT ammonium nitrate.