Vacin and went modified orchid medium with agar and sucrose
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PMV895A-10X1L PMV895B-10L PMV895C-50L
APOSPMV895A-10X1LEA 0 EUR
APOSPMV895A-10X1L APOSPMV895B-10L APOSPMV895C-50L
Vacin and went modified orchid medium with agar and sucrose
Celcultuurmedia
Contains the macro- and micronutrients as described by Vacin and Went (1949).
  • Plant tissue culture tested