Sodium perchlorate monohydrate 98,0-102,0 Reag. USP, ACS

Leverancier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Perchloric acid sodium salt

134387.1210
APPC134387.1210EA 350 EUR
APPC134387.1210
Sodium perchlorate monohydrate 98,0-102,0 Reag. USP, ACS
Sodium perchlorate monohydrate
Formule: NaClO₄·H₂O
MW: 140.46 g/mol
Smeltpunt: 130
MDL Nummer: MFCD00149164
CAS nummer: 7791-07-3
UN: 1502
ADR: 5.1 II
Merck Index: 12,08798

Order Now

Specification Test Results

Appearance White crystalline powder
Assay (Arg.) 98,0 - 102,0%
pH of 5% solution  6,0-8,0
Insoluble matter in H₂O 0,005 %
Chloride (Cl)  0,003%
Nitrogen compounds (as N) 0,001%
Phosphate (PO₄) 0,001 %
Sulfate (SO₄)  0,002%
Chlorate (ClO₃)  0,05%
Heavy metals (as Pb) 0,0005%
Calcium (Ca) 0,0005%
Cadmium (Cd) 0,0005%
Cobalt (Co) 0,0005%
Copper (Cu) 0,0005%
Iron (Fe) 0,01%
Potassium (K) 0,005%
Magnisium (Mg) 0,0005%
Manganese (Mn) 0,0005%
Nickel (Ni) 0,0005%
Lead (Pb) 0,0005%
Zinc (Zn) 0,0005%
Solubility in water 2,090 g/l at 20 °C, hygroscopic
Thermal decomposition Above 130 °C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR