Benzylalcohol 98.0-100.5% (by acidometric titration), liquid USP, NF, Ph. Eur., BP, pharma grade

Leverancier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Benzenemethanol, Phenylmethanol, Phenylcarbinol

141081.0719 141081.0816 141081.1214 141081.1611
APPC141081.0719EA 0 EUR
APPC141081.0719 APPC141081.0816 APPC141081.1214 APPC141081.1611
Benzylalcohol 98.0-100.5% (by acidometric titration), liquid USP, NF, Ph. Eur., BP, pharma grade
Benzylalcohol
Formule: C₆H₅CH₂OH
Kookpunt: 204,7 °C
Smeltpunt: –15,2 °C
Dichtheid: 1,043 – 1.049 (20°C)
Vlampunt: 94 °C
MDL Nummer: MFCD00004599
CAS nummer: 100-51-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearence Clear colourless liquid
Solubility in water Soluble in water 40 g/L at 20 °C D 20/4 1,0453
Flash point 94 °C 0,13 hPa
Vapor pressure 20 °C
Assay (acidic medium) 98,0 – 100,5%
Refractive index n20/D 1,5396
Ignition temperature 435 °C
Dielectric constant 25 °C 13,1
Heat evaporation 205 °C 467 KJ/Kg
Saturation concentration 20 °C 0,56 g/m³
Explosion limit 1,3% (V) 13% (V)
Density 1,043 – 1,049 (20/20)
Non-volatile matter 0,05%
Residue on ignition (as SO₄) 0,005%
Peroxide value 5
Benzaldehyde (G.C.) 0,15%
Ciclohexylmethanol 0,10%
Halogenated compounds and halides (as Cl) 0,03%
Water (H₂O) 0,2%
Cadmium (Cd) 0,5 ppm
Lead (Pb) 0,5 ppm
Arsenic (As) 1,5 ppm
Mercury (Hg) 1,5 ppm
Cobalt (Co) 5 ppm
Vanadium (V) 10 ppm
Nickel 20 ppm
Titanium 0,8 ppm
Gold (Au) 10 ppm
Palladium (Pd) 10 ppm
Iridium 10 ppm
Osmium (Os) 10 ppm
Rhodium (Rh) 10 ppm
Ruthenium (Ru) 10 ppm
Selenium (Se) 10 ppm
Silver (Ag) 10 ppm
Platinum (Pt) 10 ppm
Lithium (Li) 55 ppm
Antimony (Sb) 120 ppm
Barium (Ba) 140 ppm
Molybdenum 25 ppm
Copper (Cu) 250 ppm
Tin (Sn) 600 ppm
Chromium (Cr) 25 ppm
Total imp. Tr < benzyl alcohol 0,04%
Passes test Colour of solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR