Potassium dihydrogen phosphate 98.0-100.5% (by acidimetry) NF, USP, BP, Ph. Eur., pharma grade

Leverancier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate

141509.0416 141509.1210 141509.1211 141509.1214
APPC141509.0416EA 1710 EUR
APPC141509.0416 APPC141509.1210 APPC141509.1211 APPC141509.1214
Potassium dihydrogen phosphate 98.0-100.5% (by acidimetry) NF, USP, BP, Ph. Eur., pharma grade
Potassium dihydrogen phosphate
Formule: KH₂PO₄
Smeltpunt: 252 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011401
CAS nummer: 7778-77-0
EINECS: 231-913-4

Order Now

Specification Test Results

Assay (Acidimetry) 98.0-100.5%
pH (5% Solution) 4.2-4.5
Loss on drying (130 °C) 1.0%
Insoluble matter in H₂O 0.2%
Reducing substances Passes test
Chloride (Cl) 0.02%
Sulphate (SO₄) 0.03%
Residual solvents (Ph. Eur./USP) Passes test
Fluoride (F) 0.001%
Heavy metals (as Pb) 0.001%
As 0.0001%
Fe 0.001%
PB 0.00005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR