Sodium hydrogen carbonate 99.0-100.5% (by acidimetry) USP, Ph. Eur., BP, pure, Pharma Grade

Leverancier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Sodium bicarbonate

141638.0416 141638.1211 141638.1214
APPC141638.0416EA 0 EUR
APPC141638.0416 APPC141638.1211 APPC141638.1214
Sodium hydrogen carbonate 99.0-100.5% (by acidimetry) USP, Ph. Eur., BP, pure, Pharma Grade
Sodium hydrogen carbonate
Formule: NaHCO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003528
CAS nummer: 144-55-8
EINECS: 205-633-8

Order Now

Specification Test Results

Assay (Acidim.) 99.0-100.5%
Identity according to Pharmacopoeias: passes test
Maximum limit of impurities
Appearance of solution passes test
Loss on drying 0.25%
Chloride (Cl) 0.005%
Sulfur compounds (as SO4) 0.015 %
Ammonium (NH4) 0.002%
Phosphate (PO4) 0.002 %
Sulfate (SO4) 0.01%
Carbonate passes test
Residual solvents (Ph.Eur/USP) passes test
As 0.00015 %
Ca 0.01 %
Fe 0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR