ortho-Fosforzuur 10% USP-NF, BP, pure, Pharma Grade

Leverancier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Phosphoric acid

147143.0716
APPC147143.0716EA 0 EUR
APPC147143.0716
ortho-Fosforzuur 10% USP-NF, BP, pure, Pharma Grade
ortho-Fosforzuur
Formule: H₃PO₄
MW: 98 g/mol
Kookpunt: 158
Smeltpunt: 21
MDL Nummer: MFCD00011340
CAS nummer: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8 III
Merck Index: 14,07344

Order Now

Specification Test Results

Assay (Acidim.) (w/w) (Ph. Eur.) 9.5 - 10.5 %
Assay (Acidim.) (w/v) (USP-NF) 9.5 - 10.5 %
Identity according to Pharmacopoeias passes test
Appearance of solution passes test Chloride (Cl): 0.0006 %
Sulfate (SO4) (Ph. Eur.) 0.0010 %
Sulfate (SO4) (USP-NF) passes test
Precipitated substances by NH4OH passes test
Residual solvents (Ph.Eur/USP) passes test
Nitrate (NO3) passes test
Hypophosphorous and phosphorous acid passes test
Alkaline phosphates: passes test Fe 0.0006 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR