Potassium chloride 99.0-100.5% (by argentometric analysis, on dry substance) USP, Ph. Eur., BP, pharma grade

Leverancier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Potassiumchloride

191494.0416 191494.1211
APPC191494.0416EA 0 EUR
APPC191494.0416 APPC191494.1211
Potassium chloride 99.0-100.5% (by argentometric analysis, on dry substance) USP, Ph. Eur., BP, pharma grade
Potassium chloride
Formule: KCl
Kookpunt: 1420 °C
Smeltpunt: 778 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011360
CAS nummer: 7447-40-7
EINECS: 231-211-8

Order Now

Specification Test Results

Assay (Argentometric) 99.0 - 100.5%
Identity According to Pharmacopoeias Passes Test
Acidity or Alkalinity Passes Test
Loss on Drying at 105 °C ≤1.0%
Bromide (Br) ≤0.1%
Sulphate (SO₄) ≤0.03%
Iodide Passes Test
Residual Solvents (Ph.Eur./USP) Passes Test
Ba (Ph. Eur.) Passes Test
Calcium, Magnesium and Alkaline-Earth Metals (as Ca) ≤0.02%
Iodide (USP) ≤0.005%
Heavy Metals (as Pb) ≤0.001%
Al ≤0.0001%
Ba ≤0.001%
Fe ≤0.002%
NA ≤0.1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR