AQUAMETRIC titrants for volumetric Karl Fischer titrations
Leverancier: PanReac AppliChem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
285815.1612 285815.1611 285816.1611
APPC285815.1612EA 275 EUR
APPC285815.1612 APPC285815.1611 APPC285816.1611
AQUAMETRIC titrants for volumetric Karl Fischer titrations
Karl Fischer titranten en reagentia
  • Use with AQUAMETRIC solvent
1 ml of Titrant 5 corresponds to: min. 5,0 mg of H₂O (20°C) at time of manufacture.
1 ml of Titrant 2 corresponds to: min. 2,0 mg of H₂O (20°C) at time of manufacture.