Disposable Visitor Coats with Stand-up Collar
Leverancier: ASATEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PKIO-KV30W-3XL PKIO-KV30W-L PKIO-KV30W-XL PKIO-KV30W-XXL
ASATPKIO-KV30W-3XLEA 77.1 EUR
ASATPKIO-KV30W-3XL ASATPKIO-KV30W-L ASATPKIO-KV30W-XL ASATPKIO-KV30W-XXL
Disposable Visitor Coats with Stand-up Collar
Wegwerpkleding
Ideal for hygiene protection and protection against dirt without health risk.
  • Stand-up collar
  • 4-point velcro fastener
  • Elasticated cuffs
  • No pockets
Lightweight, highly breathable.