Anti-TMEM176A Rabbit Polyclonal Antibody

Leverancier: Aviva Systems Biology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ARP43539_P050
AVIVARP43539_P050EA 376 EUR
AVIVARP43539_P050
Anti-TMEM176A Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: TMEM176A (transmembrane protein 176A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR