Anti-ADAM19 Rabbit Polyclonal Antibody

Leverancier: Aviva Systems Biology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ARP49780_P050
AVIVARP49780_P050EA 376 EUR
AVIVARP49780_P050
Anti-ADAM19 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: ADAM19 (ADAM metallopeptidase domain 19)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR