Anti-C14orf177 Rabbit Polyclonal Antibody

Leverancier: Aviva Systems Biology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ARP55821_P050
AVIVARP55821_P050EA 376 EUR
AVIVARP55821_P050
Anti-C14orf177 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: C14orf177 (chromosome 14 open reading frame 177)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Specificaties

antigeen symbool C14orf177
Type antilichaam Primair,Chromosome 14 open reading frame 177
Environmentally Preferable
Type antilichaam Primair,Chromosome 14 open reading frame 177
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Human
Host Rabbit
Gen-ID 283598
Isotype
Western blot Yes
Epitoop N-terminal
Kruisadsorptie none
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR