Anti-CLEC4G Rabbit Polyclonal Antibody

Leverancier: Aviva Systems Biology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ARP55883_P050
AVIVARP55883_P050EA 376 EUR
AVIVARP55883_P050
Anti-CLEC4G Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: CLEC4G (C-type lectin domain family 4, member G)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR