Anti-RAB23 Rabbit Polyclonal Antibody

Leverancier: Aviva Systems Biology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ARP56870_P050
AVIVARP56870_P050EA 376 EUR
AVIVARP56870_P050
Anti-RAB23 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: RAB23 (RAB23, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Specificaties

antigeen symbool RAB23
Type antilichaam Primair,RAB23, member RAS oncogene family
Environmentally Preferable
Antigeen synoniemen HSPC137
Type antilichaam Primair,RAB23, member RAS oncogene family
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity
Host Rabbit
Gen-ID 51715
Isotype
Western blot Yes
Epitoop N-terminal
Kruisadsorptie none
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR