Anti-CAB39 Rabbit Polyclonal Antibody

Leverancier: Aviva Systems Biology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ARP56877_P050
AVIVARP56877_P050EA 376 EUR
AVIVARP56877_P050
Anti-CAB39 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: CAB39 (calcium binding protein 39)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR