Anti-CCT2 Rabbit Polyclonal Antibody

Leverancier: Aviva Systems Biology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ARP63402_P050
AVIVARP63402_P050EA 376 EUR
AVIVARP63402_P050
Anti-CCT2 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: CCT2 (chaperonin containing TCP1, subunit 2 (beta))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Specificaties

antigeen symbool CCT2
Environmentally Preferable
Type antilichaam Primair,chaperonin containing TCP1, subunit 2 (beta)
Antigeen synoniemen TCP-1-beta|PRO1633|CCTB|99D8.1|HEL-S-100n|CCT-beta
Type antilichaam Primair,chaperonin containing TCP1, subunit 2 (beta)
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity
Host Rabbit
Gen-ID 10576
Isotype
Western blot Yes
Epitoop N-terminal
Kruisadsorptie none
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR