Anti-RORA Rabbit Polyclonal Antibody

Leverancier: Aviva Systems Biology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

P100794_T100
AVIVP100794_T100EA 327 EUR
AVIVP100794_T100
Anti-RORA Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: RORA (RAR-related orphan receptor A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Specificaties

antigeen symbool RORA
Environmentally Preferable
Type antilichaam Primair,RAR-related orphan receptor A
Antigeen synoniemen RZRA|RZR-ALPHA|ROR3|ROR1|NR1F1|ROR2
Type antilichaam Primair,RAR-related orphan receptor A
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Human
Host Rabbit
Gen-ID 6095
Isotype
Western blot Yes
Epitoop N-terminal
Kruisadsorptie none
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR