Hand- en vingerbeschermers, Azlon®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PGR406
AZLOPGR406EA 59.7 EUR
AZLOPGR406
Hand- en vingerbeschermers, Azlon®
Handbeschermers
Hand protector made of silicone rubber.
  • Ideal for safe handling of hot beakers, flasks etc
  • Capable of withstanding temperatures up to 250 °C
  • Also excellent for handling cold objects down to −57 °C
  • Contact surfaces are multi-studded for positive grip
* One size fits all.