Sleeves for ground joints, Azlon®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SWN961 SWN962 SWN963 SWN964 SWN965 SWN966 SWN968 SWN969
AZLOSWN961EA 41.3 EUR
AZLOSWN961 AZLOSWN962 AZLOSWN963 AZLOSWN964 AZLOSWN965 AZLOSWN966 AZLOSWN968 AZLOSWN969
Sleeves for ground joints, Azlon®
Geslepen Geslepen hulzen
Sleeves, PTFE, for conical ground joints.
  • Chemical resistance
  • Allows easy separation of joints
  • Intended for single or occasional use
  • Forms a gas tight seal
  • No contamination from joint grease
Working temperature: −100 to +250 °C.
Certificaten: Suitable for food contact applications in accordance with USA regulations, USP Class VI grade PTFE.