4-Bromo-2-ethyl-1-iodobenzene ≥97% stabilised

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


Stabilisatie: Stabilised with copper

B20052.09 B20052.88 B20052.36
B20052.09EA 166 EUR
B20052.09 B20052.88 B20052.36
4-Bromo-2-ethyl-1-iodobenzene ≥97% stabilised
4-Bromo-2-ethyl-1-iodobenzene

Stabilized with copper


Purity: 97% CAS No.: 175278-30-5 Molecular Formula: C8H8BrI Formula Weight: 310.97 Boiling Point: 84-86°/0.4mm MDL No.: MFCD00221458

Formule: C₈H₈BrI
Kookpunt: 84…86 °C (0,4 mmHg)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00221458
CAS nummer: 175278-30-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR