5-Bromoindole ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B20307.06 B20307.14 B20307.22
B20307.06EA 29.3 EUR
B20307.06 B20307.14 B20307.22
5-Bromoindole ≥99%
5-Bromoindole

Purity: 99% CAS No.: 10075-50-0 Molecular Formula: C8H6BrN Formula Weight: 196.05 Odour: Odourless Melting Point: 89-92° Beilstein Reference: 112877 MDL No.: MFCD00005670

Formule: C₈H₆BrN
Smeltpunt: 89…92 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005670
CAS nummer: 10075-50-0
EINECS: 233-208-7

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR