N-Methylanthranilic acid ≥90%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B20531.14 B20531.22 B20531.36 AB125880.0100
B20531.14EA 33.9 EUR
B20531.14 B20531.22 B20531.36 ABCRAB125880.0100
N-Methylanthranilic acid ≥90%
N-Methylanthranilic acid

Purity: 90+% CAS No.: 119-68-6 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Melting Point: ca 171° dec. Beilstein Reference: 2208135

Formule: C₈H₉NO₂
Kookpunt: 80 °C (0,01 mmHg)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002424
CAS nummer: 119-68-6
EINECS: 204-343-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR