α-Bromo-p-toluonitrile ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

B20641.06 B20641.14
B20641.06EA 46.9 EUR
B20641.06 B20641.14
α-Bromo-p-toluonitrile ≥98%
α-Bromo-p-toluonitrile

Purity: 98% CAS No.: 17201-43-3 Molecular Formula: C8H6BrN Formula Weight: 196.05 Melting Point: 113-117° Beilstein Reference: 636717 UN No.: 2923 MDL No.: MFCD00001829

Formule: C₈H₆BrN
Kookpunt: 135…137 °C (0,6 mmHg)
Smeltpunt: 113…117 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001829
CAS nummer: 17201-43-3
EINECS: 241-246-0
UN: 2923
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR